missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 10763831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763831 100 g Plastic bottle 100g
10763641 5 g Glass Bottle 5g
10627452 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10763831 Supplier Thermo Scientific Acros Supplier No. 111941000
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 520-45-6 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-45-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00066709
InChI Key PGRHXDWITVMQBC-UHFFFAOYSA-N
Synonym dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
PubChem CID 122903
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
SMILES CC1=CC(=O)C(C(=O)O1)C(=O)C

Specifications

Melting Point 109.0°C to 112.0°C
Color Cream or Yellow
Boiling Point 270.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 17, 559
Merck Index 15, 2869
Solubility Information Solubility in water: <0.1% (25 c). Other solubilities: 22g/100g acetone- 18g/100g benzene (25 c),5g/100g methanol- 3g/100g ccl4 (25 c),3g/100g ethanol- 0.7g/100g n-heptane (25 c),1.7g/100g propylene glycol- 5g/100g ether (25 c)
Formula Weight 168.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Dehydroacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Dehydroacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.