missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydrodolichyl Diphosphate Synthase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | Dehydrodolichyl Diphosphate Synthase |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Dehydrodolichyl Diphosphate Synthase Polyclonal antibody specifically detects Dehydrodolichyl Diphosphate Synthase in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Dehydrodolichyl Diphosphate Synthase | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cis-isoprenyltransferase, cis-prenyl transferase, CIT, CPT, Dedol-PP synthase, dehydrodolichyl diphosphate synthase, EC 2.5.1.-, FLJ13102, HDSDS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human DHDDS (NP_995583.1). IKAGPMPKHIAFIMDGNRRYAKKCQVERQEGHSQGFNKLAETLRWCLNLGILEVTVYAFSIENFKRSKSEVDGLMDLARQKFSRLMEEKEKLQKHGVCIRVLGDLHLLPLDLQELIAQAVQATKNYNKCFLNVCFAYTSRHEISNAVREMAWGVEQGLLDPSDISESLLDKCLYTNRSPHP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 79947 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title