Learn More
Thermo Scientific™ Deionizer Cartridges
Product Code. 10071893 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Milli-Q Millipore Water Systems
Nanopure
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Mixed Bed, Ultrapure, Oxygen and Organic Removal
Oxygen Removal Mixed Bed
Oxygen Removal Resin
Porous Strong Anion Exchange
Porous Strong Ion Exchange
Strong Anion Exchange
Strong Cation Exchange
Super-C Carbon
Two Bed Ion Exchange
Ultrapure, Oxygen Removal
Ultrapure, Oxygen and Organic Removal
Capacity:
1680 grains
1760 grains
18 g
2000 gal.
275 grains/2000 gal.
275 grains/9 g
30 g
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
5000 gal.
5000 gal. (as CaCO3)
550 grains
785 grains/1000 gal.
875 grains
Unit Size:
1 unit
Each
For Use With (Application):
Anion Removal
Cation Removal
High capacity, two-bed ion exchange
Ion Exchange, Ultrapure
Organic Removal
Oxygen Removal
Oxygen and demineralization
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure and Oxygen Removal Resin
Ultrapure, Oxygen and Organic Removal
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Resin Type Capacity Unit Size For Use With (Application)
10071893 Bantam™ Deionizer, B-Pure™ Mixed Bed 875 grains Each -
12740666 Bantam™ Deionizer, B-Pure™ Strong Anion Exchange 1680 grains Each Anion Removal
14871651 Nanopure Mixed Bed - Each Ultrapure
15342178 B-Pure™ Activated Carbon 5000 gal. (as CaCO3) 1 unit Organic Removal
15352178 B-Pure™ Mixed Bed - 1 unit Ultrapure
15362178 B-Pure™ Oxygen Removal Resin - 1 unit Oxygen Removal
12331683 B-Pure™ Ultrapure, Oxygen Removal - Each Ultrapure, Oxygen and Organic Removal
15372178 B-Pure™ Mixed Bed, Ultrapure, Oxygen and Organic Removal - 1 unit Ultrapure and Oxygen Removal Resin
11789428 Milli-Q Millipore Water Systems Super-C Carbon - Each Organic Removal
11799428 Milli-Q Millipore Water Systems Mixed Bed - Each Ion Exchange, Ultrapure
10589690 B-Pure™ Porous Strong Ion Exchange 30 g Each Oxygen Removal
10041953 Bantam™ Deionizer, B-Pure™ Activated Carbon 5000 gal. Each Organic Removal
12311278 Bantam™ Deionizer, B-Pure™ Strong Cation Exchange 3170 grains Each Cation Removal
11746189 Bantam™ Deionizer, B-Pure™ Mixed Bed, Activated Carbon 785 grains/1000 gal. Each Ultrapure and Organic Removal
10451820 B-Pure™ Macroreticular, Activated Carbon 5000 gal. Each Pretreatment
10297093 B-Pure™ Macroreticular, Activated Carbon 2000 gal. Each Pretreatment Macropure
10074067 B-Pure™ Mixed Bed 550 grains Each Ultrapure
15312168 B-Pure™ Porous Strong Anion Exchange 18 g 1 unit Oxygen Removal
15322168 B-Pure™ Activated Carbon 5000 gal. 1 unit Organic Removal
15297643 B-Pure™ Oxygen Removal Mixed Bed 275 grains/9 g Each Oxygen and demineralization
10125767 B-Pure™ Mixed Bed, Activated Carbon 275 grains/2000 gal. Each Ultrapure and Organic Removal
11706289 B-Pure™ Extruded Carbon 5000 gal. Each Organic Removal
12730746 B-Pure™ Ultrapure, Oxygen Removal 455 grains/30 g Each Ultrapure and Organic Removal
10081893 B-Pure™ Ultrapure, Oxygen and Organic Removal 365 grains/2000 gal./12 g Each Ultrapure, Oxygen and Organic Removal
10501673 Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated Two Bed Ion Exchange 1760 grains Each High capacity, two-bed ion exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Product Code. 10071893 Supplier Thermo Scientific™ Supplier No. D0809
Wastewater Testing Savings
Use promo code 28023
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
TRUSTED_SUSTAINABILITY
Specifications
Capacity 875 grains
Resin Type Mixed Bed
For Use With (Equipment) Bantam™ Deionizer, B-Pure™
Type Deionizer Cartridge