missing translation for 'onlineSavingsMsg'
Learn More

Delanzomib, MedChemExpress

Product Code. 16484986
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484986

Brand: Medchem Express HY10454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Delanzomib (CEP-18770) is a potent and orally active chymotrypsin-like activity of the proteasome inhibitor with an IC50 of 3.8 nM. Delanzomib inhibits NF-κB activity, induces cancer cell apoptotic, and has strong antiangiogenic and anti-cancer activities.

Delanzomib (CEP-18770) is a potent and orally active chymotrypsin-like activity of the proteasome inhibitor with an IC50 of 3.8 nM. Delanzomib inhibits NF-κB activity, induces cancer cell apoptotic, and has strong antiangiogenic and anti-cancer activities.

Specifications

Chemical Name or Material Delanzomib
CAS 847499-27-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H28BN3O5
Quantity 100 mg
Synonym CEP-18770
Solubility Information DMSO : 100 mg/mL (241.97 mM; Need ultrasonic)
SMILES CC(C)C[C@@H](B(O)O)NC([C@@H](NC(C1=NC(C2=CC=CC=C2)=CC=C1)=O)[C@H](O)C)=O
Molecular Weight (g/mol) 413.28
Formula Weight 413.28
Percent Purity 96.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.