Learn More
Lenz Laborglasintrumente Delivery Adapters Straight with Suction Tube
Product Code. 10131811
Change view
Click to view available options
Standard Taper Joint No.:
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. unitSize
10131811 NS 14/23 Each
10599741 NS 29/32 Each
10589741 NS 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10131811 Supplier Lenz Laborglasintrumente Supplier No. 05266001
  • ONLINE EXCLUSIVE PRICE
    27.00 €
    22.15 € / Pack of 1
    Save 4.85 €
    17% Off
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Delivery adapters
  • Straight
  • With suction tube
  • With NS socket and NS cone
  • With drain tip
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. NS 14/23