Learn More
Lenz Laborglasintrumente Delivery Adapters Straight with Suction Tube
Product Code. 10599741
Change view
Click to view available options
Standard Taper Joint No.:
NS 14/23
NS 19/26
NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. Unit Size
10599741 NS 29/32 Each
10131811 NS 14/23 Each
10589741 NS 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10599741 Supplier Lenz Laborglasintrumente Supplier No. 05266004
  • ONLINE EXCLUSIVE PRICE
    37.00 €
    23.90 € / Pack of 1
    Save 13.10 €
    35% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Delivery adapters
  • Straight
  • With suction tube
  • With NS socket and NS cone
  • With drain tip
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. NS 29/32