Learn More
Cole-Parmer™ Delrin™ Manual Pinch Valve
Specifications
Specifications
| Material | Delrin |
| For Use With | 5/32 in. to 1/4 in. Tube O.D. |
| For Use With (Application) | Ideal for use in pharmaceutical and food industries where eliminating contamination is critical |
| Packaging Quantity | 1 Pack |
| Product Type | Manual Pinch Valve |