Learn More
Chemsplash AG MAX Type 4B/5B/6B Coverall
GREENER_CHOICE
Product Code. 30058231
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058231
Brand: Chemsplash 2658.043.004
Only null left
Add to basket
Add to basket
AG MAX is made from a 65 gsm microporous laminated Cat III Type 4B, 5B and 6B fabric.
Chemsplash AG MAX Coverall
  • 65 GSM Dark Green Microporous Laminate Fabric
  • Fully Taped Seams
  • Three Piece Hood
  • Elasticated Cuffs with Thumb Loops
  • Elasticated Hood, Back and Ankles
  • Two Way Zip with Zip Flap
  • Silicone and Latex Free
  • Low Linting
  • Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
  • Protects Against Pesticides ISO 27065: 2017 Class 2
Description: The Chemsplash AG MAX is made from a 65GSM microporous laminated Cat III Type 4B, 5B and 6B fabric. This coverall offers enhanced protection against pesticides, in accordance with ISO 27065: 2017 Class 2. All seams are taped to provide a stronger, more effective barrier against liquids and particulate, ensuring maximum protection for users.
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Composite Fabric
Color Green
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)