Learn More
Chemsplash Delta Coverall (Sterile Irradiated)
Product Code. 30058292
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058292 Large Pack of 25
30058449 Small Pack of 25
30058369 Medium Pack of 25
30058401 X-Large Pack of 25
30058175 2X-Large Pack of 25
30058230 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058292 Supplier Chemsplash Supplier No. 2766.400.006
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    149.09 € / Pack of 25
    Save 1.91 €
    1% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Delta 67 Coverall - Sterile Irradiated - is made from our soft comfortable 67 gsm microporous laminate fabric, meeting Cat III Type 5B/6B.
Chemsplash Delta 67 Coverall
  • 67GSM Microporous Laminate Fabric
  • Sterile Irradiated, Double Bagged and Detex Sticker
  • Low Linting Knitted Cuffs with Thumb Loops
  • Elasticated Three Piece Hood
  • Two Way Zip
  • Adhesive Zip Flap
  • Elasticated Back and Ankles
  • Reinforced Bound Seam at Crotch Area
  • Non Linting and Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Coverall is made from soft, comfortable 67gsm microporous laminate fabric, meeting Cat III Type 5B/6B. It is Sterile Irradiated, double-bagged, and comes with a Detex sticker for identification. The coverall is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). Chemsplash Delta is ideal for prolonged wear, offering improved breathability while maintaining an effective hazard barrier. This coverall is suitable for a wide range of applications, with an elasticated back, hood, cuffs, ankles, and reinforced seams for added durability. It conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9.
Specifications
Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit, Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)