Learn More
Chemsplash Jet Spray 88 Type 3/4 5/6 Coverall
GREENER_CHOICE
Product Code. 30058402
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058402
Brand: Chemsplash 2727.700.007
Only null left
Add to basket
Add to basket
Jet Spray 88 Type 3B/4B 5B/6B Coverall is highly resistant against Chemical permeation offering high levels of protection against even the most corrosive of chemicals.
Chemsplash Jet Spray 88 Type 3B/4B 5B/6B Coverall
  • 88GSM Heavyweight Microporous Non-Woven Fabric
  • Ultrasonically Tape Welded Seams
  • Self Adhesive Chin Strap for Optimum Protection
  • Elasticated Hood, Half Waist, Cuffs, and Ankles
  • Convenient Thumb Loops at Wrist
  • Single-Way Zip with Bi-Folding Self Adhesive Flap
  • Silicone and Latex Free
  • Non Linting Fabric
  • Anti-Static
  • PFAS Free
Description: The Chemsplash Jet Spray 88 Type 3B/4B 5B/6B Coverall is designed to provide high resistance against chemical permeation, offering exceptional protection against even the most corrosive chemicals. Made from a durable 88gsm heavy-weight Polypropylene/Polyethylene multilayer laminated material, it also provides protection against biological hazards, including viruses, bacteria, and blood-borne pathogens, according to EN14126. In addition, the coverall offers protection against particulate radioactive contamination (level 2) as per EN1073-2 standards. The design includes a self-adhesive chin strap, elasticated hood, waist, cuffs, and ankles, and thumb loops for increased comfort and protection. This coverall is perfect for environments where high levels of chemical and biological protection are required.
Specifications
Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material Microporous Composite Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)