missing translation for 'onlineSavingsMsg'
Learn More

delta-Decanolactone, 97%

Product Code. 11499303
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499303 100 g 100g
11489303 25 g 25g
11409313 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499303 Supplier Thermo Scientific Alfa Aesar Supplier No. A11756.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 705-86-2 | C10H18O2 | 170.25 g/mol

Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

Solubility
Soluble in water (4 mg/ml at 28°C), alcohols, and propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-86-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00006649
InChI Key GHBSPIPJMLAMEP-UHFFFAOYNA-N
Synonym delta-decalactone, 6-pentyltetrahydro-2h-pyran-2-one, 5-decanolide, decanolide-1,5, 5-decalactone, decan-5-olide, amyl-delta-valerolactone, delta-decanolactone, 5-pentyl-5-pentanolide, 2h-pyran-2-one, tetrahydro-6-pentyl
PubChem CID 12810
ChEBI CHEBI:87327
IUPAC Name 6-pentyloxan-2-one
SMILES CCCCCC1CCCC(=O)O1

Specifications

Melting Point -25°C
Density 0.971
Boiling Point 117°C to 120°C (0.02 mmHg)
Flash Point 145°C (293°F)
Refractive Index 1.458
Quantity 100 g
Beilstein 117520
Solubility Information Soluble in water (4mg/ml at 28°C),alcohols,and propylene glycol.
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material delta-Decanolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.