missing translation for 'onlineSavingsMsg'
Learn More

delta-Hexanolactone, 98%

Product Code. 11469536
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469536 100 g 100g
11459536 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469536 Supplier Thermo Scientific Alfa Aesar Supplier No. A19274.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-22-3 | C6H10O2 | 114.144 g/mol

d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-22-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00083574
InChI Key RZTOWFMDBDPERY-UHFFFAOYSA-N
Synonym delta-hexalactone, delta-hexanolactone, delta-caprolactone, 5-methyl-delta-valerolactone, 5-hydroxyhexanoic acid lactone, tetrahydro-6-methyl-2h-pyran-2-one, 2h-pyran-2-one, tetrahydro-6-methyl, .delta.-caprolactone, 6-methylvalerolactone, hexanolactone
PubChem CID 13204
IUPAC Name 6-methyloxan-2-one
SMILES CC1CCCC(=O)O1

Specifications

Melting Point 18°C
Density 1.04
Boiling Point 231°C
Flash Point 105°C (221°F)
Refractive Index 1.451
Quantity 100 g
Beilstein 80501
Solubility Information Immiscible with water.
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material delta-Hexanolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.