missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactone, 98%, may contain polymer

Product Code. 11301717
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301717 25 g 25g
11311717 100 g 100g
11321717 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11301717 Supplier Thermo Scientific Alfa Aesar Supplier No. L04794.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 542-28-9 | C5H8O2 | 100.12 g/mol

δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
δ-Valerolactone is employed as a chemical intermediate in the synthesis of polymers. It is also used to synthesize the copolyesters by lipase-catalyzed ring opening polymerization.

Solubility
Miscible with water.

Notes
Store in a tightly closed container. Avoid breathing dust, vapor, mist, or gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-28-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00006645
InChI Key OZJPLYNZGCXSJM-UHFFFAOYSA-N
Synonym delta-valerolactone, tetrahydro-2h-pyran-2-one, 5-valerolactone, 2h-pyran-2-one, tetrahydro, tetrahydropyran-2-one, tetrahydro-2-pyranone, penta-1,5-lactone, .delta.-valerolactone, .delta.-valeryllactone, tetrahydro-pyran-2-one
PubChem CID 10953
ChEBI CHEBI:16545
IUPAC Name oxan-2-one
SMILES O=C1CCCCO1

Specifications

Melting Point -13°C to -12°C
Density 1.056
Boiling Point 58°C to 60°C (0.5 mmHg)
Flash Point 112°C (233°F)
Refractive Index 1.458
Quantity 25 g
Beilstein 106436
Solubility Information Miscible with water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material delta-Valerolactone, may contain polymer; distil to remove
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.