missing translation for 'onlineSavingsMsg'
Learn More

Demecolcine, TRC

Product Code. 30008482
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008482 1 mg 1mg
30004444 10 mg 10mg
30003581 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008482 Supplier Toronto Research Chemicals Supplier No. TRCD2307851MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-30-5
Molecular Formula C21 H25 N O5
Molecular Weight (g/mol) 371.43
Synonym Colcemid, Benzo[a]heptalen-9(5H)-one, 6,7-dihydro-1,2,3,10-tetramethoxy-7-(methylamino)-, (S)-, Colchamine, Colchicine, N-deacetyl-N-methyl-, Colchicine, N-deacetyl-N-methyl-, (-)-, (-)-Colchamine, (-)-Demecolcine, Alkaloid H 3, from Colchicum antumnale, C 12669, Ciba 12669A, Colchamin, Deacetyl-N-methylcolchicine, Deacetylmethylcolchicine, Demecolcin, Demecolcine, Desacetylmethylcolchicine, Desmecolcine, Kolchamin, N-Deacetyl-N-methylcolchicine, N-Desacetyl-N-methylcolchicine, N-Methyl-N-deacetylcolchicine, N-Methyl-N-desacetylcolchicine, N-Methyltri-O-methylcolchicinic acid, NSC 3096, Omain, Omaine, Reichstein's F, Santavy's substance F, Substance F
IUPAC Name (7S)-1,2,3,10-tetramethoxy-7-(methylamino)-6,7-dihydro-5H-benzo[a]heptalen-9-one
SMILES CN[C@H]1CCc2cc(OC)c(OC)c(OC)c2C3=CC=C(OC)C(=O)C=C13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H25NO5/c1-22-15-8-6-12-10-18(25-3)20(26-4)21(27-5)19(12)13-7-9-17(24-2)16(23)11-14(13)15/h7,9-11,15,22H,6,8H2,1-5H3/t15-/m0/s1
Formula Weight 371.1733
Chemical Name or Material Demecolcine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.