missing translation for 'onlineSavingsMsg'
Learn More

Demethyl Benzydamine Hydrochloride, TRC

Product Code. 30135187
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135187 5 mg 5mg
30132480 2.5 mg 2.5mg
17912135 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135187 Supplier Toronto Research Chemicals Supplier No. TRCD2308105MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39860-97-4
Molecular Formula C18H22ClN3O
Molecular Weight (g/mol) 331.84
Synonym N-Methyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine Hydrochloride, 1-Benzyl-3-(3-methylaminopropoxy)-1H-indazole Monohydrochloride;
SMILES CNCCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H21N3O.ClH/c1-19-12-7-13-22-18-16-10-5-6-11-17(16)21(20-18)14-15-8-3-2-4-9-15;/h2-6,8-11,19H,7,12-14H2,1H3;1H
Chemical Name or Material Demethyl Benzydamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.