missing translation for 'onlineSavingsMsg'
Learn More

Demeton-S-methyl Sulfoxide, TRC

Product Code. 30040451
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040451 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040451 Supplier Toronto Research Chemicals Supplier No. TRCD2313802.5MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 301-12-2
Molecular Formula C6 H15 O4 P S2
Quantity 2.5 mg
Synonym Ethanethiol, 2-(ethylsulfinyl)-, S-ester with O,O-dimethyl phosphorothioate (8CI), BAY 21097, Demeton-S methyl sulfoxide, MSR, MSR (pesticide), MSR Spray Concentrate, Metaisosystox sulfoxide, Metasystox R, Methyl oxydemeton S, NSC 370785, O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl] monothiophosphate, O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl] phosphorothioate, O,O-Dimethyl S-[2-(ethylsulfinyl)ethyl] thiophosphate, Oxydemeton M, Oxydemeton-methyl, R 2170, S-[2-(Ethylsulfinyl)ethyl] O,O-dimethyl phosphorothioate
InChI Formula InChI=1S/C6H15O4PS2/c1-4-13(8)6-5-12-11(7,9-2)10-3/h4-6H2,1-3H3
SMILES CCS(=O)CCSP(=O)(OC)OC
IUPAC Name 1-dimethoxyphosphorylsulfanyl-2-ethylsulfinylethane
Molecular Weight (g/mol) 246.28
Formula Weight 246.0149
Chemical Name or Material Demeton-S-methyl sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.