missing translation for 'onlineSavingsMsg'
Learn More

Denatonium benzoate

Product Code. 15493709
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493709 1 g 1g
15403719 5 g 5g
15413719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493709 Supplier Thermo Scientific Alfa Aesar Supplier No. J61048.03
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The bitterest compound known | CAS: 3734-33-6 | C28H34N2O3

The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.

Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Synonym unii-m5ba6gaf1o

Specifications

Quantity 1 g
Solubility Soluble in isopropyl alcohol,methanol. Slightly soluble in acetone. Insoluble in ether.
Melting Point 164°C to 169°C
Formula Weight 446.59
Physical Form Solid
Color White
Chemical Name or Material Denatonium benzoate
compliance-icons
Product Identifier
  • Denatonium benzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bitrex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.