missing translation for 'onlineSavingsMsg'
Learn More

Denatonium Benzoate, TRC

Product Code. 30008934
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008934 25 g 25g
30009700 1 g 1g
30004946 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008934 Supplier Toronto Research Chemicals Supplier No. TRCD23166025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C21 H29 N2 O . C7 H5 O2
Molecular Weight (g/mol) 446.58
Synonym Denatonium Benzoate, Ammonium, benzyldiethyl[(2,6-xylylcarbamoyl)methyl]-, benzoate (8CI), Benzenemethanaminium, N-[2-[(2,6-dimethylphenyl)amino]-2-oxoethyl]-N,N-diethyl-, benzoate (9CI), Benzyldiethyl[(2,6-xylylcarbamoyl)methyl]ammonium benzoate (7CI), Anispray, Aversion, Bitrex, Bitterant Bitrex PG, Denatonium benzoate, ECX 95BY, Gori, Lignocaine benzyl benzoate, N-[2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]-N,N-diethyl(phenylmethyl)ammonium benzoate, NSC 157658, THS 839, Win 16568
IUPAC Name benzyl-[2-(2,6-dimethylanilino)-2-oxoethyl]-diethylazanium;benzoate
SMILES CC[N+](CC)(CC(=O)Nc1c(C)cccc1C)Cc2ccccc2.[O-]C(=O)c3ccccc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C21H28N2O.C7H6O2/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;8-7(9)6-4-2-1-3-5-6/h7-14H,5-6,15-16H2,1-4H3;1-5H,(H,8,9)
Formula Weight 446.2569
Percent Purity >95%
Chemical Name or Material Bitrex (TM)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.