missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Denaturing Lysis Buffer

Product Code. 15442959
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
Product Code. Quantity unitSize
15442959 250mL 250mL
15452959 500mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 15442959

Brand: Thermo Scientific™ J60132.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
pH 7.4
Content And Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.