missing translation for 'onlineSavingsMsg'
Learn More

Dendrobine, TRC

Product Code. 30364955
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364955 100 mg 100mg
30366327 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30364955 Supplier Toronto Research Chemicals Supplier No. TRCD231445100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2115-91-5
Molecular Formula C16H25NO2
Molecular Weight (g/mol) 263.38
Synonym Dendroban-12-one, (-)-Dendrobine, 12-Oxodendrobane, [2aS-(2aα,4aα,5β,6α,7β,7aα,7bα)]-Decahydro-1,7b-dimethyl-6-(1-methylethyl)-7,5-(epoxymethano)-1H-cyclopent[cd]indol-9-one, NSC 607862
SMILES Cl.CCNC1(CCCCC1)c2cc(OC)ccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H25NO2/c1-8(2)11-12-10-6-5-9-7-17(4)14(16(9,10)3)13(11)19-15(12)18/h8-14H,5-7H2,1-4H3/t9-,10+,11+,12-,13-,14-,16+/m1/s1
Chemical Name or Material Dendrobine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.