missing translation for 'onlineSavingsMsg'
Learn More

Deoxy Artemisinin, TRC

Product Code. 30361281
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361281 10 mg 10mg
30358083 100 mg 100mg
30355378 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361281 Supplier Toronto Research Chemicals Supplier No. TRCD23215010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72826-63-2
Molecular Formula C15 H22 O4
Molecular Weight (g/mol) 266.33
Synonym 10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, octahydro-3,6,9-trimethyl-, (3R,3aS,6R,6aS,9S,10aS,10bR)-, 10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, octahydro-3,6,9-trimethyl-, [3R-(3α,3aβ,6β,6aβ,9β,10aα,10bβ)]-, (3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, 2-Deoxyartemisinin, 2-Desoxyartemisinin, Deoxyarteannuin, Deoxyartemisinin, Deoxyqinghaosu, Desoxyartemisinin, Hydroarteannuin, Qing Hau Sau III, Qinghaosu III, (3R,3aS,3a1R,6R,6aS,9S,10aS)-3,6,9-trimethyldecahydro-2H-3a1,9-epoxyoxepino[4,3,2-ij]isochromen-2-one
SMILES C[C@@H]1CC[C@H]2[C@@H](C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23O4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H22O4/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-15/h8-11,13H,4-7H2,1-3H3/t8-,9-,10+,11+,13-,14-,15-/m1/s1
Formula Weight 266.1518
Chemical Name or Material Deoxy Artemisinin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.