missing translation for 'onlineSavingsMsg'
Learn More

Deoxy Corticosterone Acetate, TRC

Product Code. 30356493
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356493 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356493 Supplier Toronto Research Chemicals Supplier No. TRCD2325955G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deoxycorticosterone acetate
CAS 56-47-3
Molecular Formula C23 H32 O4
Quantity 5 g
Synonym Desoxycortone acetate, 3,20-Dioxopregn-4-en-21-yl acetate, 11-Deoxycorticosterone, acetate, Pregn-4-ene-3,20-dione, 21-hydroxy-, acetate, 11-Deoxycorticosterone 21-acetate, 11-Desoxycorticosterone acetate, 21-Acetoxy-3,20-diketopregn-4-ene, 21-Acetoxypregn-4-en-3,20-dione, 21-Acetoxypregn-4-ene-3,20-dione, 21-Hydroxypregn-4-ene-3,20-dione 21-acetate, 21-Hydroxypregn-4-ene-3,20-dione acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnene-3,20-dione 21-acetate, 4-Pregnene-3,20-dione-21-ol acetate, Arcort, Bio-Corten, Cortacet, Cortate, Cortenil, Cortesan, Cortexone acetate, Cortifar, Cortigen, Cortinaq, Cortiron, Cortivis, Cortixyl, DCA, DOC acetate, DOCA, Decorten, Decortin, Decosteron, Decosterone, Decostrate, Deoxycorticosterone 21-acetate, Deoxycorticosterone acetate, Descorterone, Descotone, Desoxycorticosterone acetate, Desoxycortone acetate, Dorcostrin, Doxo, Doxycamon, Krinocorts, NSC 9567, Ocriten, Ocritena, Percorten, Percotol, Primocort, Primocortan, Sincortex, Steraq, Syncort, Syncorta, Syncortyl, Unidocan
InChI Formula InChI=1S/C23H32O4/c1-14(24)27-13-21(26)20-7-6-18-17-5-4-15-12-16(25)8-10-22(15,2)19(17)9-11-23(18,20)3/h12,17-20H,4-11,13H2,1-3H3/t17-,18-,19-,20+,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [2-[(8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 372.5
Formula Weight 372.2301
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.