missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Deoxycholic Acid, Sodium Salt

Product Code. 11486292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486292 25 g 25g
11496292 100 g 100g
11406302 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486292 Supplier MP Biomedicals™ Supplier No. 0210290625
  • ONLINE EXCLUSIVE PRICE
    52.05 €
    49.35 € / 25g
    Save 2.70 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Deoxycholic Acid, Sodium Salt is an anionic detergent used to solubilize membrane-bound proteins.

  • Deoxycholic acid, sodium salt is an anionic detergent useful for the extraction of membrane receptors and other plasma membrane proteins and for nuclei isolation.
  • Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/mL solution.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
CAS 302-95-4
Molecular Formula C24H39NaO4
Formula Weight 414.55
MDL Number MFCD00064139
Form Powder
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Molecular Weight (g/mol) 414.56
Quantity 25 g
Show More Show Less
compliance-icons
Product Identifier
  • Deoxycholic Acid Sodium Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: deoxycholic acid sodium salt

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.