missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Deoxycholic Acid, Sodium Salt

Product Code. 11496292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496292 100 g 100g
11486292 25 g 25g
11406302 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496292 Supplier MP Biomedicals™ Supplier No. 0210290680
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    177.45 € / 100g
    Save 10.55 €
    5% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Deoxycholic Acid, Sodium Salt is an anionic detergent used to solubilize membrane-bound proteins.

  • Deoxycholic acid, sodium salt is an anionic detergent useful for the extraction of membrane receptors and other plasma membrane proteins and for nuclei isolation.
  • Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/mL solution.

Specifikationer

Color White
Product Type Detergent
CAS 302-95-4
Molecular Formula C24H39NaO4
Formula Weight 414.55
Form Powder
Quantity 100 g
compliance-icons
Produktbeteckning
  • Deoxycholic Acid Sodium Salt Monohydrate
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contain: sodium 3-alpha, 12-alpha-dihydroxy-5-holan-24-oate

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.