missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Deoxycholic acid, 98.5%

Product Code. 10616732 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616732 25 g Glass bottle 25g
10796181 100 g Glass bottle 100g
10636904 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616732 Supplier Thermo Scientific Chemicals Supplier No. 148140250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

Deoxycholic acid, CAS # 83-44-3, is a bile acid and blood serum metabolite. | CAS: 83-44-3 | C24H40O4 | 392.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Deoxycholic acid is a 5beta-cholan-24-oic acid derivative with hydroxy substitutions at carbons 3 and 12

• Deoxycholic acid’s role is to absorb fats by emulsifying and solubilizing them

Applications

• Deoxycholic acid may be used for laboratory study of submental fat

• Deoxycholic acid is capable of disrupting cell membranes

• In laboratory research applications, deoxycholic acid can be used to solubilize proteins or as a secondary standard

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifica

Melting Point 172°C to 178°C
Color Pink to White
Ignition Residue 0.2 % max.
Loss on Drying 1 % max. (140°C, 4 hrs) (under vacuum < 5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Quantity 25 g
Beilstein 10, IV, 1608
Merck Index 15, 2901
Specific Rotation +53° to +57° (20°C, 589nm) (c=1, EtOH)
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Formula Weight 392.57
Percent Purity 98.0 to 102.0% (on dried basis) (Acid)
Physical Form Powder
Chemical Name or Material Deoxycholic acid
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Deoxycholic acid
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.