missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Deoxycholic acid, 98.5%

Product Code. 10796181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10796181 100 g Glass bottle 100g
10616732 25 g Glass bottle 25g
10636904 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10796181 Supplier Thermo Scientific Chemicals Supplier No. 148141000
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Deoxycholic acid, CAS # 83-44-3, is a bile acid and blood serum metabolite. | CAS: 83-44-3 | C24H40O4 | 392.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Deoxycholic acid is a 5beta-cholan-24-oic acid derivative with hydroxy substitutions at carbons 3 and 12

• Deoxycholic acid’s role is to absorb fats by emulsifying and solubilizing them

Applications

• Deoxycholic acid may be used for laboratory study of submental fat

• Deoxycholic acid is capable of disrupting cell membranes

• In laboratory research applications, deoxycholic acid can be used to solubilize proteins or as a secondary standard

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 172°C to 178°C
Color Pink to White
Ignition Residue 0.2 % max.
Loss on Drying 1 % max. (140°C, 4 hrs) (under vacuum < 5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Quantity 100 g
Beilstein 10, IV, 1608
Merck Index 15, 2901
Specific Rotation +53° to +57° (20°C, 589nm) (c=1, EtOH)
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Formula Weight 392.57
Percent Purity 98.0 to 102.0% (on dried basis) (Acid)
Physical Form Powder
Chemical Name or Material Deoxycholic acid
Show More Show Less
compliance-icons
Product Identifier
  • Deoxycholic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.