missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid, >99%, MP Biomedicals™

Product Code. 17871361 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17871361 10 g 10g
17881361 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17871361 Supplier MP Biomedicals Supplier No. 0210149610
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
IUPAC Name (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color White
Assay Percent Range >99%
Quantity 10 g
Formula Weight 392.6
Physical Form Crystalline Powder
Chemical Name or Material Deoxycholic acid
compliance-icons
Product Identifier
  • Deoxycholic Acid, free acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3-alpha,12-alpha-dihydroxy-5-ß-cholan-24-oic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.