missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid sodium salt, MedChemExpress

Product Code. 16499106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499106 100 mg 100mg
16432386 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499106 Supplier Medchem Express Supplier No. HYN0593A
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Deoxycholic acid sodium salt (sodium deoxycholate), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Deoxycholic acid sodium salt (sodium deoxycholate), a bile acid, is a by-product of intestinal metabolism, that activates the G protein-coupled bile acid receptorTGR5.

Specifications

Chemical Name or Material Deoxycholic acid sodium salt
CAS 302-95-4
Color White
Purity Grade Notes Research
Molecular Formula C24H39NaO4
Quantity 100 mg
Synonym Sodium deoxycholate
Solubility Information H2O : 333.33 mg/mL (804.08 mM; Need ultrasonic) ∣DMSO : 6.25 mg/mL (15.08 mM; Need ultrasonic)
SMILES C[C@@]1([C@@]2([H])[C@H](C)CCC(O[Na])=O)[C@](CC2)([H])[C@@](CC[C@@]3([H])[C@@]4(CC[C@@H](O)C3)C)([H])[C@]4([H])C[C@@H]1O
Molecular Weight (g/mol) 414.55
Formula Weight 414.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.