missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic Acid, TRC

Product Code. 30007481
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007481 10 g 10g
30005826 25 g 25g
17951585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007481 Supplier Toronto Research Chemicals Supplier No. TRCD23264510G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24 H40 O4
Molecular Weight (g/mol) 392.57
Synonym Cholan-24-oic acid, 3,12-dihydroxy-, (3α,5β,12α)-, 5β-Cholan-24-oic acid, 3α,12α-dihydroxy- (8CI), (3α,5β,12α)-3,12-Dihydroxycholan-24-oic acid, 17β-[1-Methyl-3-carboxypropyl]-etiocholane-3α,12α-diol, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholanoic acid, 3α,12α-Dihydroxycholanic acid, 5β-Cholanic acid-3α,12α-diol, 5β-Deoxycholic acid, 7-Deoxycholic acid, ATX 101, Cholerebic, Cholic acid, deoxy-, Cholorebic, Degalol, Deoxycholatic acid, Deoxycholic acid, Desoxycholic acid, Droxolan, Kybella, NSC 8797, Pyrochol, Septochol
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1
Formula Weight 392.2927
Chemical Name or Material Deoxycholic Acid (Choleic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.