missing translation for 'onlineSavingsMsg'
Learn More

Deoxynivalenol, TRC

Product Code. 17909759
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909759 10 mg 10mg
18023450 5 mg 5mg
18071940 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17909759 Supplier Toronto Research Chemicals Supplier No. TRCD24390010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51481-10-8
Molecular Formula C15 H20 O6
Molecular Weight (g/mol) 296.32
Synonym (3α,7α)-12,13-Epoxy-3,7,15-trihydroxytrichothec-9-en-8-one, Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane], trichothec-9-en-8-one deriv., 4-Deoxynivalenol, 4-Desoxynivalenol, DON, Dehydronivalenol, Deoxynivalenol, NSC 269144, Vomitoxin
SMILES CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1
Formula Weight 296.126
Chemical Name or Material Deoxynivalenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.