missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic acid, Sodium Salt (not highly polymerized), 91%, MP Biomedicals™

Product Code. 11418531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418531 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418531 Supplier MP Biomedicals Supplier No. 0215227525
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Deoxyribonucleic acid sodium salt has been used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin

Specifications

CAS 9007-49-2
Color Cream
Assay Percent Range 91%
Molecular Formula C15H31N3O13P2
MDL Number MFCD00130921
Quantity 25 g
Synonym nucleic acid, na salt yeast, deoxyribonucleic acid herring sperm , not higly polimerized, 5-amino-2-5-amino-2-hydroxymethyl oxolan-3-yl oxy hydroxy phosphoryl oxy methyl oxolan-3-yl oxy 5-amino-3-hydroxyoxolan-2-yl methoxyphosphinic acid
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
SMILES *
IUPAC Name [5-amino-2-[[[5-amino-2-(hydroxymethyl)oxolan-3-yl]oxy-hydroxyphosphoryl]oxymethyl]oxolan-3-yl] (5-amino-3-hydroxyoxolan-2-yl)methyl hydrogen phosphate
Molecular Weight (g/mol) 0.00
PubChem CID 44135672
Physical Form Powder
Chemical Name or Material Deoxyribonucleic acid, Sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.