missing translation for 'onlineSavingsMsg'
Learn More

Melford DEPC

Product Code. 30018238
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018238 5 mL 5mL
30018237 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018238 Supplier Melford Supplier No. D430605.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Modification reagent for His and Tyr residues in proteins.

Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.

Specifications

CAS 1609-47-8
Molecular Formula C6H10O5
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.