missing translation for 'onlineSavingsMsg'
Learn More

Melford DEPC

Product Code. p-200087234
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 30018237

Brand: Melford D4306025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Modification reagent for His and Tyr residues in proteins.

Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.

Specifications

CAS 1609-47-8
Molecular Formula C6H10O5
Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.