missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deprodone propionate, MedChemExpress
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Deprodone propionate (RD20000) is a corticosteroid which is obtained by esterifying with propionic acid the 17-position of the prednisolone skeleton and deoxidating its 21-position.
Specifications
Specifications
| Chemical Name or Material | Deprodone propionate |
| CAS | 20424-00-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H32O5 |
| Quantity | 10 mg |
| Synonym | RD20000 |
| Solubility Information | DMSO : 250 mg/mL (624.20 mM; Need ultrasonic) |
| SMILES | C[C@@]12[C@@](C(C)=O)(CC[C@@]1([H])[C@@]3([H])[C@]([C@@]4(C(CC3)=CC(C=C4)=O)C)([H])[C@@H](O)C2)OC(CC)=O |
| Molecular Weight (g/mol) | 400.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction