missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Dermanios Scrub CG

Product Code. 11776555
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11776555 1 L Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11776555 Supplier Laboratoires Anios™ Supplier No. 2082523

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antiseptic soap for hygienic handwashing.

Antiseptic Soap:

  • Clear solution, slightly viscous
  • Product pH : approx. 5
  • Density : approx. 1.0

Specifications

Product Type Dermanios Scrub
Composition Digluconate de chlorhexidine excipients
For Use With (Application) For Surgical Environment, Medical and Industrial Clean Room
Quantity 12Pk.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.