missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ DermaShield™ 73-711 Series Green Neoprene Gloves
BIOPHARMAGREENER_CHOICE

Product Code. 12842193
Change view
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Unit Size:
200 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12842193 7.5 200 pairs
12475070 6.5 200 pairs
12852193 8.5 200 pairs
12425080 9 200 pairs
12405080 8 200 pairs
12485070 7 200 pairs
12465070 6 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12842193 Supplier Ansell™ Supplier No. 73711075
Add to basket
Edge
Add to basket

Most technically advanced sterile glove available today. Free of latex proteins and accelerators, it minimizes both Type I and Type IV allergies associated with natural rubber.

  • Made of poly-chloroprene
  • Free of latex proteins and accelerators, it minimises both Type I and Type IV allergies associated with natural rubber.
  • Comfort and flexibility of natural rubber gloves at a similarly competitive price
  • Neoprene composition offers better chemical resistance.
  • New beaded cuff prevents the glove from rolling down, providing increased protection.
  • Sterile
  • Available for non-sterile controlled environments.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Neoprene
Cuff Material Neoprene
Applications Laboratory
Color Green
Latex Free Yes
Material Neoprene
Powdered Powder Free
Product Type Neoprene Gloves
Size 7.5
Sterility Sterile
Thickness (Metric) 0.175 mm
Glove Cuff Color Green
Glove Cuff Style Beaded/Rolled
Quantity 200/CS
Resistant To Chemical resistance
Series 73-711
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.