missing translation for 'onlineSavingsMsg'
Learn More

Dermatan Sulfate (>90%), TRC

Product Code. 30135096
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135096 25 mg 25mg
30007818 10 mg 10mg
30010193 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135096 Supplier Toronto Research Chemicals Supplier No. TRCD28932525MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54328-33-5
Synonym Chondroitin Sulfate B Sodium Salt, Chondroitin Sulfate Type B Sodium Salt, Dermatan Sulfate Sodium Salt;
IUPAC Name sodium (3S,6R)-6-(((2S,5S)-3-acetamido-2-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)tetrahydro-2H-pyran-4-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H23NO15S.2Na/c1-3(17)15-5-10(9(30-31(24,25)26)4(2-16)27-13(5)23)28-14-8(20)6(18)7(19)11(29-14)12(21)22;;/h4-11,13-14,16,18-20,23H,2H2,1H3,(H,15,17)(H,21,22)(H,24,25,26);;/q;2*+1/p-2/t4?,5?,6?,7-,8?,9+,10?,11?,13-,14+;;/m0../s1
Chemical Name or Material Chondroitin sulfate B sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.