missing translation for 'onlineSavingsMsg'
Learn More

Des[5-(2-dimethylamino)ethyl] Diltiazem, TRC

Product Code. 30366646
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366646 50 mg 50mg
30366348 10 mg 10mg
30358380 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366646 Supplier Toronto Research Chemicals Supplier No. TRCD28895050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87447-47-0
Molecular Formula C18 H17 N O4 S
Molecular Weight (g/mol) 343.4
Synonym (2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate, Diltiazem Hydrochloride Imp. B (EP)
IUPAC Name [(2S,3S)-2-(4-methoxyphenyl)-4-oxo-3,5-dihydro-2H-1,5-benzothiazepin-3-yl] acetate
SMILES COc1ccc(cc1)[C@@H]2Sc3ccccc3NC(=O)[C@@H]2OC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H17NO4S/c1-11(20)23-16-17(12-7-9-13(22-2)10-8-12)24-15-6-4-3-5-14(15)19-18(16)21/h3-10,16-17H,1-2H3,(H,19,21)/t16-,17+/m1/s1
Formula Weight 343.0878
Chemical Name or Material (2S,3S)-2-(4-Methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.