missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Diltiazem Hydrochloride, TRC

Product Code. 30008897
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008897 100 mg 100mg
30010949 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008897 Supplier Toronto Research Chemicals Supplier No. TRCD289335100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75472-91-2
Molecular Formula C20 H24 N2 O3 S . Cl H
Molecular Weight (g/mol) 408.94
Synonym 1,5-Benzothiazepin-4(5H)-one, 5-[2-(dimethylamino)ethyl]-2,3-dihydro-3-hydroxy-2-(4-methoxyphenyl)-, hydrochloride (1:1), (2S,3S)-, 1,5-Benzothiazepin-4(5H)-one, 5-[2-(dimethylamino)ethyl]-2,3-dihydro-3-hydroxy-2-(4-methoxyphenyl)-, monohydrochloride, (2S,3S)- (9CI), 1,5-Benzothiazepin-4(5H)-one, 5-[2-(dimethylamino)ethyl]-2,3-dihydro-3-hydroxy-2-(4-methoxyphenyl)-, monohydrochloride, (2S-cis)-, Deacetyldiltiazem hydrochloride
IUPAC Name (2S,3S)-5-[2-(dimethylamino)ethyl]-3-hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4-one;hydrochloride
SMILES Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24N2O3S.ClH/c1-21(2)12-13-22-16-6-4-5-7-17(16)26-19(18(23)20(22)24)14-8-10-15(25-3)11-9-14;/h4-11,18-19,23H,12-13H2,1-3H3;1H/t18-,19+;/m1./s1
Formula Weight 408.1274
Percent Purity >95%
Chemical Name or Material Desacetyl Diltiazem HCl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.