missing translation for 'onlineSavingsMsg'
Learn More

Deschloro Amlodipine, TRC

Product Code. 30358068
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358068 10 mg 10mg
30351148 2.5 mg 2.5mg
30353252 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358068 Supplier Toronto Research Chemicals Supplier No. TRCD28977510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88150-52-1
Molecular Formula C20 H26 N2 O5
Molecular Weight (g/mol) 374.43
Synonym 2-[(2-Aminoethoxy)methyl]-1,4-dihydro-6-methyl-4-phenyl-3,5-pyridinedicarboxylic Acid 3-Ethyl 5-Methyl Ester
IUPAC Name 3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-6-methyl-4-phenyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2)C(=O)OC)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H26N2O5/c1-4-27-20(24)18-15(12-26-11-10-21)22-13(2)16(19(23)25-3)17(18)14-8-6-5-7-9-14/h5-9,17,22H,4,10-12,21H2,1-3H3
Formula Weight 374.1842
Chemical Name or Material Deschloro Amlodipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.