missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descyano Cimetidine Dihydrochloride Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride |
| CAS | 59660-24-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 H17 N5 S . 2 Cl H |
| Quantity | 100 mg |
| Synonym | 1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride, Cimetidine Hydrochloride Imp. D (EP) as Dihydrochloride |
| InChI Formula | InChI=1S/C9H17N5S.2ClH/c1-7-8(14-6-13-7)5-15-4-3-12-9(10)11-2;;/h6H,3-5H2,1-2H3,(H,13,14)(H3,10,11,12);2*1H |
| SMILES | Cl.Cl.CNC(=N)NCCSCc1nc[nH]c1C |
| IUPAC Name | 1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]guanidine;dihydrochloride |
| Molecular Weight (g/mol) | 300.25 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?