missing translation for 'onlineSavingsMsg'
Learn More

Desipramine-2,4,6,8-d4 Hydrochloride, TRC

Product Code. 30357498
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357498 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30357498 Supplier Toronto Research Chemicals Supplier No. TRCD2900521MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61361-34-0
Molecular Formula C18 2H4 H18 N2 . Cl H
Quantity 1 mg
Synonym 5H-Dibenz[b,f]azepine-2,4,6,8-d4-5-propanamine, 10,11-dihydro-N-methyl-, monohydrochloride (9CI), Desipramine-2,4,6,8-D4 Hydrochloride
InChI Formula InChI=1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H/i2D,3D,9D,10D;
SMILES Cl.[2H]c1cc([2H])c2N(CCCNC)c3c([2H])cc([2H])cc3CCc2c1
IUPAC Name N-methyl-3-(1,3,8,10-tetradeuterio-5,6-dihydrobenzo[b][1]benzazepin-11-yl)propan-1-amine;hydrochloride
Molecular Weight (g/mol) 306.87
Formula Weight 306.1801
Chemical Name or Material Desipramine-2,4,6,8-d4 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.