missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Desipramine hydrochloride

Product Code. 18792484
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792484 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18792484 Supplier R&D Systems™ Supplier No. 3067/50
  • ONLINE EXCLUSIVE PRICE
    145.00 €
    136.50 € / 50mg
    Save 8.50 €
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of noradrenalin transporters

Tricyclic antidepressant that is a selective inhibitor of noradrenalin transporters (Ki values are 4, 61 and 78720 nM for NET, SERT and DAT transporters respectively).
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-28-6
Quantity 50 mg
Product Line Tocris
Molecular Formula C18H23ClN2
Formula Weight 302.84
Purity >99%
Molecular Weight (g/mol) 302.846
Chemical Name or Material Desipramine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.