missing translation for 'onlineSavingsMsg'
Learn More

Desipramine Hydrochloride, TRC

Product Code. 30132601
Change view
Click to view available options
Quantity:
5 mg
100 mg
1 g
Unit Size:
100mg
1g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132601 1 g 1g
30136814 100 mg 100mg
18084030 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132601 Supplier Toronto Research Chemicals Supplier No. TRCD2900501G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-28-6
Molecular Formula C18 H22 N2 . Cl H
Molecular Weight (g/mol) 302.84
Synonym Desipramine Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N-methylpropan-1-amine Hydrochloride, Imipramine Hydrochloride Imp. A (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N-methyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 10,11-dihydro-5-[3-(methylamino)propyl]-, monohydrochloride (8CI), 5H-Dibenz[b,f]azepine-5-propanamine, 10,11-dihydro-N-methyl-, monohydrochloride (9CI), 10,11-Dihydro-5-[3-(methylamino)propyl]-5H-dibenz[b,f]azepine monohydrochloride, 5-[3-(Methylamino)propyl]-5H-dibenz[b,f]azepine hydrochloride, DMI hydrochloride, Demethylimipramine hydrochloride, Desimipramine-hydrochloride, Desipramine monohydrochloride, Desmethylimipramine chloride, Desmethylimipramine monohydrochloride, Desmethylimipramine-hydrochloride, EX 4355, G 35020, GMI, JB 8181, N-(γ-Methylaminopropyl)iminodibenzyl hydrochloride, NSC 114901, Norpramin, Nortrimil, Pertofran, Pertofrane, RMI 9384A
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N-methylpropan-1-amine;hydrochloride
SMILES Cl.CNCCCN1c2ccccc2CCc3ccccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19H,6,11-14H2,1H3;1H
Formula Weight 302.155
Chemical Name or Material Desipramine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.