missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desisobutyryl Ciclesonide-d11 (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | Desisobutyryl Ciclesonide cyclohexyl-d11 (mixture of diastereomers) |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 2H11 H27 O6 |
| Quantity | 10 mg |
| Synonym | (11β,16α)-16,17-[[(R/S)-Cyclohexyl-d11-methylene]bis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, CIC-AP-d11, Ciclesonide active principle-d11 |
| InChI Formula | InChI=1S/C28H38O6/c1-26-11-10-18(30)12-17(26)8-9-19-20-13-23-28(22(32)15-29,27(20,2)14-21(31)24(19)26)34-25(33-23)16-6-4-3-5-7-16/h10-12,16,19-21,23-25,29,31H,3-9,13-15H2,1-2H3/t19-,20-,21-,23+,24+,25?,26-,27-,28+/m0/s1/i3D2,4D2,5D2,6D2,7D2,16D |
| SMILES | [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C2O[C@@H]3C[C@H]4[C@@H]5CCC6=CC(=O)C=C[C@]6(C)[C@H]5[C@@H](O)C[C@]4(C)[C@@]3(O2)C(=O)CO)C([2H])([2H])C1([2H])[2H] |
| Molecular Weight (g/mol) | 481.67 |
| Formula Weight | 481.3359 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?