missing translation for 'onlineSavingsMsg'
Learn More

SymLine Table Gland for Desktop Mount

Product Code. 10150040
Change view
Click to view available options
Closure Size:
G 1/2 in. ID (f), 33.5 mm OD on GL 45 (m)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
10150040 G 1/2 in. ID (f), 33.5 mm OD on GL 45 (m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150040 Supplier SymLine Supplier No. 106400
  • ONLINE EXCLUSIVE PRICE
    144.00 €
    120.38 € / Pack of 1
    Save 23.62 €
    16% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Desktop mount accessory with G 1/2 in. ID (f), 33.5 mm OD on GL 45 (m)

SymLine is the intelligent, modular collection system for liquid waste. It integrates perfectly into your laboratory equipment. It was specially developed for the requirements in the analytical laboratory.

Specifications

Closure Size G 1/2 in. ID (f), 33.5 mm OD on GL 45 (m)
Description Desktop mount accessory with G 1/2 in. ID (f), 33.5 mm OD on GL 45 (m), PE-HD electrostatic conductive incl. gasket and lock nμt
Quantity 1 Ea.
For Use With (Application) Transfer of Liquids Into Connected Waste Systems
For Use With (Equipment) Can be Combined with 3 x and 4 x Collector, SafetyWasteCap, Safety Funnel and Pipe System
Material HDPE
Product Type Desktop Mount
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.