missing translation for 'onlineSavingsMsg'
Learn More

Desloratadine N-Hydroxypiperidine, TRC

Product Code. 30137866
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137866 50 mg 50mg
30133955 25 mg 25mg
30011225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137866 Supplier Toronto Research Chemicals Supplier No. TRCD29026550MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193725-73-3
Molecular Formula C19 H19 Cl N2 O
Molecular Weight (g/mol) 326.82
Synonym 8-Chloro-6,11-dihydro-11-(1-hydroxy-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, 4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidin-1-ol
IUPAC Name 8-chloro-11-(1-hydroxypiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES ON1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H19ClN2O/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-21-19(14)18(17)13-7-10-22(23)11-8-13/h1-2,5-6,9,12,23H,3-4,7-8,10-11H2
Formula Weight 326.1186
Chemical Name or Material Desloratadine N-Hydroxypiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.