missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Norflurazon, TRC

Product Code. 30011029
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011029 250 mg 250mg
17991965 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011029 Supplier Toronto Research Chemicals Supplier No. TRCD296865250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23576-24-1
Molecular Formula C11 H7 Cl F3 N3 O
Molecular Weight (g/mol) 289.64
Synonym 3(2H)-Pyridazinone, 5-amino-4-chloro-2-[3-(trifluoromethyl)phenyl]-, 3(2H)-Pyridazinone, 5-amino-4-chloro-2-(α,α,α-trifluoro-m-tolyl)- (8CI), 5-Amino-4-chloro-2-[3-(trifluoromethyl)phenyl]-3(2H)-pyridazinone, 4-Chloro-5-amino-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone, Desmethylnorflurazon, SAN 9774, Norflurazon-desmethyl
IUPAC Name 5-amino-4-chloro-2-[3-(trifluoromethyl)phenyl]pyridazin-3-one
SMILES NC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H7ClF3N3O/c12-9-8(16)5-17-18(10(9)19)7-3-1-2-6(4-7)11(13,14)15/h1-5H,16H2
Formula Weight 289.023
Percent Purity >95%
Chemical Name or Material Norflurazon-desmethyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.