missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl PBR28, TRC

Product Code. 30359340
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359340 10 mg 10mg
30353452 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359340 Supplier Toronto Research Chemicals Supplier No. TRCD29244510MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005325-42-7
Molecular Formula C20H18N2O3
Molecular Weight (g/mol) 334.37
Synonym Desmethyl PBR28, N-[(2-Hydroxyphenyl)methyl]-N-(4-phenoxy-3-pyridinyl)acetamide
SMILES CC(=O)N(Cc1ccccc1O)c2cnccc2Oc3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H18N2O3/c1-15(23)22(14-16-7-5-6-10-19(16)24)18-13-21-12-11-20(18)25-17-8-3-2-4-9-17/h2-13,24H,14H2,1H3
Formula Weight 334.1317
Chemical Name or Material N-[(2-Hydroxyphenyl)methyl]-N-(4-phenoxy-3-pyridinyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.