missing translation for 'onlineSavingsMsg'
Learn More

Desonide 21-Acetate, TRC

Product Code. 30358744
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358744 1 g 1g
30350355 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358744 Supplier Toronto Research Chemicals Supplier No. TRCD2969251G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25092-25-5
Molecular Formula C26 H34 O7
Molecular Weight (g/mol) 458.54
Synonym Acetyldesonide, 11β-Hydroxy-16α,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxopregna-1,4-dien-21-yl acetate, Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-acetate (7CI,8CI), (11β,16α)-21-(Acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Desonide 21-acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H34O7/c1-14(27)31-13-20(30)26-21(32-23(2,3)33-26)11-18-17-7-6-15-10-16(28)8-9-24(15,4)22(17)19(29)12-25(18,26)5/h8-10,17-19,21-22,29H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,22+,24-,25-,26+/m0/s1
Formula Weight 458.2305
Percent Purity >95%
Chemical Name or Material Desonide 21-Acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.